Would you like to log in again?
Diamond Sharpening Stones, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62082-51 62082-52 62082-50
100497-606EA 82.79 USD InStock
100497-606 100497-608 100497-610
Diamond Sharpening Stones, Electron Microscopy Sciences
Scissors
The patented heat treatment process bonds the highest quality industrial grade diamond particles in a stainless alloy to a precision matched metal substrate. It has been imitated, but has never come close to being duplicated.
  • Sharpening stone set on a walnut pedestal with a cover
  • Available in fine grit (600), medium grit (270), and coarse grit (180)
  • Stone size is 2 x 6" (51 x 152 mm)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty