Would you like to log in again?
Specimen Mailers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61003 61004 61011 61015
100489-736PK 210.26 USD InStock
100489-736 100489-734 100489-744 100489-740
Specimen Mailers, Electron Microscopy Sciences
Mailers and Shippers
The Double Mailing Container set consists of an inner aluminum screw cap container and an outer fiberboard screw cap mailing tube.
These fiberboard screw cap mailing cases meet all US Postal Regulations in mailing biological specimens. Also meet IATA regulations regarding shipments of infectious substances. They are constructed of heavy duty fiberboard and have crimped metal rims and seamed metal bottoms. The metal screw caps are autoclavable and offer a secure closure. Available in several sizes.
The Double Mailing Container set consists of an inner aluminum screw cap container and an outer fiberboard screw cap mailing tube. This double type unit is required by US Postal for mailing of etiologic agents or biohazard specimens. Each aluminum container is autoclavable and the cap is lined to prevent leakage.
This double type unit is required by US Postal for mailing of etiologic agents or biohazard specimens. Each aluminum container is autoclavable and the cap is lined to prevent leakage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty