Would you like to log in again?
Tempfix Adhesive, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
12668
100505-020EA 93.08 USD InStock
100505-020
Tempfix Adhesive, Electron Microscopy Sciences
Stains Microscopy and staining reagents
A thermoplastic adhesive for mounting powder specimens and small particles for SEM.
  • Does not contain any solvents
  • Stable in high vacuum
  • Not sticky at room temperature
Adhesive does not contain any solvents and it is stable in high vacuum
Quality Standards: ISO 9001; EN29001; ANSI/ASQC Q91.
It is not sticky at room temperature but becomes adhesive at 40°C and melts at 120°C. Tempfix may also be used as an embedding medium. EMS introduces a new reliable line of conductive adhesives which are certified by the Bureau Veritas Quality International.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty