Would you like to log in again?
Terg-A-Zyme® with Protease Enzyme, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Terg-A-Zyme®
60521 60520
100504-776CS 782.83 USD InStock
100504-776 100504-832
Terg-A-Zyme® with Protease Enzyme, Electron Microscopy Sciences
Cleaning Agents Detergents and Surface Cleansers
Combine Alconox powder with this protease enzyme to effectively remove proteinaceous soils.
Ideal for pre-soaking. It also cleans reverse osmosis installations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty