Would you like to log in again?
The Cell Culture Mold, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70167
100491-606EA 51.2 USD InStock
100491-606
The Cell Culture Mold, Electron Microscopy Sciences
Moulds Moulds for Histology
This mold is designed primarily for embedding cell culture specimens grown either directly on the bottom of a dish or on the filter when cross sections of the cells are needed
Many cells can be examined in cross section. There is no need for trimming - sectioning can begin immediately. The curved side makes the tip very strong in spite of its thinness. Easy detection of a strip of cells. The label is placed on the round part so as not to interfere with the specimen. The mold has 24 cavities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty