Would you like to log in again?
The Electrode Flexible Arm, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64366-01 64366-02
100498-702EA 252.71 USD InStock
100498-702 100498-704
The Electrode Flexible Arm, Electron Microscopy Sciences
Electrode Holders
This flexible arm assists by holding and positioning electrodes or other objects in an direction. The base is solid polyethylene and comes complete with 3-M double sided mounting tape on the bottom to secure the unit to the table top
The unit is available in either a one or two hole head configuration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty