Would you like to log in again?
Tissue Embedding Center, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62300
100496-156EA 29358.35 USD InStock
100496-156
Tissue Embedding Center, Electron Microscopy Sciences
Tissue Embedders
The new tissue embedding center is equipped with the latest in circuit board and component technology.
  • Uniquely designed with microprocessor temperature sensors
  • Single console which maximizes space utilization
  • High-contrast viewing areas and multifunctional work stations
The entire unit is encased in a durable, non-heat conductive housing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty