Would you like to log in again?
Technovit® 3040 - Mounting Medium, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Technovit®
14652
100503-466KT 285.82 USD InStock
100503-466
Technovit® 3040 - Mounting Medium, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Technovit 3040 yellow is a fast curing methyl methacrylate-based resin, whose chemical composition warrants a firm, durable bond between Technovit and the specimen. Technovit® 3040 consists of two components – powder and liquid – allowing simple mixing, easy adherance to the specimen, and fast curing. For fixing the mount, a high viscous consistency is required(i.e. a mixing ratio of approximately 2-3 parts per volume powder: 1 part per volume liquid) has proven to be the most advantageous. Technovit® 3040 Kit consists of: 1 x 100g Technovit® 3040 powder and 1 x 80 mL Technovit™ 3040 liquid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty