Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tween® 20 (Polysorbate), Aurion

Supplier: Electron Microscopy Sciences
Synonyms: Polyoxyethylene sorbitan monolaurate

25564
100487-942EA 41.87 USD InStock
100487-942
Tween® 20 (Polysorbate), Aurion
Tween® 20 (Polysorbate)
TWEEN-20® is a non-ionic surfactant with hydrophobic alkyl side chains.

In an aqueous environment the components form micelles above the critical micel concentration (at approximately 0.07% in PBS). This is based on hydrophobic interaction. When other hydrophobic components (proteins, gold conjugates etc.) are present these components are likely to get embedded in micellar structures. This has several effects: hydrophobic (background) interactions arelargely suppressed, the added different molecules are slightly increased in size and previously existing hydrophobic complexes are broken up due to surface forces.


Formula: C₅₈H₁₁₄O₂₆
MW: 1227.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty