Would you like to log in again?
Vibration Foepple Dot, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68038-25
100499-296EA 490.22 USD InStock
100499-296
Vibration Foepple Dot, Electron Microscopy Sciences
Graticules Reticles and Grids
The pair of dots, which appear to merge into a single dot, determines the amount of vibration of the slide in the appropriate axis. The pattern on the S25 is an array of 20 pairs of dots converging on a single dot. The distance between each dot pair increases by 0.001" to a maximum of 0.02", pairs being squispaced 0.25".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty