Would you like to log in again?
Yankee Film Squeegee, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74822
100492-258EA 15.38 USD InStock
100492-258
Yankee Film Squeegee, Electron Microscopy Sciences
Specimen Mounting Supplies
Made from rugged high impact styrene with a 4" long and 1" wide cellulose sponge.
Easy to handle and it accommodates all films up to 4 x 5".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty