Would you like to log in again?
TIPCHECK for SPM and AFM, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
80130-TI
100495-374EA 459.55 USD InStock
100495-374
TIPCHECK for SPM and AFM, Electron Microscopy Sciences
Services
The Nioprobe device is used to determine the shape at the very apex of the tip probe for microscopy measurements.
The Problem: The physical probe used in AFM imaging is not ideally sharp. As a consequence, an AFM image does not reflect the true sample topography impartially, but rather represents the interaction of the tip with the sample surface. There is no avoiding this imperfection, which sets real limits on what may be validly inferred from an AFM image. Whether one is engaged in detailed, quantitative metrology or is simply using AFM images as a interpretive aid, it is imperative be able to assess these limits. The key here is to possess a reliable estimate of the sharpness of the tip apex. Reverse imaging of the probe is the most convenient means of obtaining the effective radius of the probe. For this purpose, the ideal characterization sample would consist of small, stiff, spiked features.
Advantages of NioProbe include the surface structure of the NioProbe film is densely populated by tiny peaks. This makes the film very suitable for the small piezo movements characteristic of precision AFM work, feature peaks exhibit imaging radii of less than 5 nm, as sharp as anything else available. This permits one to obtain the accurate apex radius desired for medium- to small- scale work (such as biomolecular imaging), the random orientation of the NioProbe features are suitable for applying blind tip reconstruction methods, the sample is resistant to the duress of contact mode scanning, the film is supplied on a silicon chip, ready to be placed in your AFM. Instructions are provided to allow easy determination of the apex radius. If stored in a clean, dry place, the sample can provide years of service.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty