Would you like to log in again?
Gold Seal Adhesion Slides , Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63734-10 63734-01
100498-566EA 1157.51 USD InStock
100498-566 100498-564
Gold Seal Adhesion Slides , Electron Microscopy Sciences
Slides Adhesive Slides
UltraStick™ (adhesion coating: APTES, 3-Aminopropyl Triethoxysilane)– Adhesion coated micro slides- one of the best from GOLD SEAL® are ideal for histological and cytological applications demanding superior specimen-to-slide adhesion.
  • Made from pure white glass (sodalime)
  • Accurately cut sizes
  • Hermetically sealed in our Moisture-Barrier™ system, providing a dry-slide environments during shipment and storage
  • Preserve the highest quality from manufacturer to ultimate user
The enhanced silane coating eliminates tedious procedures and special additives to your water bath. Procedures such as immunoperoxidase, in situ DNA hybridization, enzyme digestion and frozen tissue processing will benefit from the amplified adhesion characteristics of this product. Overall cost savings and assured quality of your prepared specimen will be recognized with UltraStick™ from Gold Seal®.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty