Would you like to log in again?
In-Situ Mount, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
17988-30
100496-558EA 138.75 USD InStock
100496-558
In-Situ Mount, Electron Microscopy Science
Stains Microscopy and staining reagents
In-Situ mounting medium is a unique mounting medium designed for permanent preservation of tissue sections and cell smear with alkaline phosphatase chromogens like NBT/BCIP and red stain. It is ideal for mounting tissues and cell smears.
In-Situ mounting medium is a unique mounting medium designed for permanent preservation of tissue sections and cell smear with alkaline phosphatase chromogens like NBT/BCIP and red stain. It is ideal for mounting tissues and cell smears. It is a direct substitute for BioMeda™ Situ Mount
*The name BioMeda™ Situ Mount is owned by BioMeda Corporation
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty