Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ERL 4221-An ERL-4206 Replacement, Electron Microscopy Science

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

15004 15005
102090-978EA 46.63 USD InStock
102090-978 102090-980
ERL 4221-An ERL-4206 Replacement, Electron Microscopy Science
Stains Microscopy and staining reagents
Due to the supply shortage of ERL-4206, ERL-4221 has now been specially formulated to replace it. ERL 4206 is one of the main components of the low viscosity (Dr. Spur) embedding resin kit.

  • Specific gravity (H2O=1) is 1.173 at 20 °C/20 °C
  • Flash point - closed cup is 118 °C (245 °F)
  • Boiling Point (760 mm/Hg) is >250 °C (>482 °F)

ERL-4221 is a Cycloaliphatic Epoxide Resin. Due to the supply shortage of ERL-4206 ERL-4221 has now been specially formulated to replace it. ERL 4206 is one of the main components of the low viscosity (Dr. Spur) embedding resin kit.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty