Would you like to log in again?
Graphite Spray Adhesive, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
12648
100504-844EA 39.43 USD InStock
100504-844
Graphite Spray Adhesive, Electron Microscopy Sciences
Stains Microscopy and staining reagents
An easy to use graphite spray to coat small samples.
  • Easy to use
  • Coats small samples
  • Fast drying
Spray adhesive is fast drying and produces a very flat, thin, and uniform graphite film. Its service temperature is up to 204°C. Its electrical resistance is 1.2kohm/sq" at 1mL thickness.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty