Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Gach (Glutaraldehyde/Carbohydrazide), Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
15920
100503-484KT 60.55 USD InStock
100503-484
Gach (Glutaraldehyde/Carbohydrazide), Electron Microscopy Sciences
Stains Microscopy and staining reagents
Hechman, C.A
A water and lipid-retaining embedding polymer for EM. Excellent preservation of lipids and ultrastructure. Kit consists of: 10 x 10 mL 50% EM grade Glutaraldehyde and 10 x 1.5 g Carbohydrazide.
Hechman, C.A. et al. (1973) J. Ultrastruct. Res. 42,156.
et al. (1973) J. Ultrastruct. Res. 42156
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty