Would you like to log in again?
Ilford Multigrade RC Cooltone Paper, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
83030-25 83030-1C
101381-636PK 73.73 USD InStock
101381-636 101381-634
Ilford Multigrade RC Cooltone Paper, Electron Microscopy Sciences
Film Processing Supplies
This is a direct replacement for Kodak Polycontrast IV RC B&W Papers.Premium Quality, Cool Tone – Variable contrast. MULTIGRADE RC COOLTONE Paper has the standard weight of (190g/m 2) with a resin coated base. It is equally suitable for printing all black and white negatives including chromogenic negatives, such as ILFORD XP2 SUPER.
  • Cooler Image Tone with Cool White Base Tint
  • Non-Developer Incorporated
  • Easy to Tone in a Variety of Toners
  • Glassy Surface
  • Developer: MULTIGRADE, or BROMOPHEN, or PQ UNIVERSAL
  • Stop Bath : ILFOSTOP, or ILFOSTOP PRO
  • Fixer: ILFORD RAPID FIXER or HYPAM
  • For ILFORD processor: ILFORD 2000RT developer/replenisher & fixer/replenisher are recommended
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty