Would you like to log in again?
Ilford 2000RT Machine Developer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
83056-05
101381-658EA 180.32 USD InStock
101381-658
Ilford 2000RT Machine Developer, Electron Microscopy Sciences
Film Processing Supplies
The 2000RT is a liquid concentrate developer/replenisher recommended for use with all types of black and white RC photo paper processors. It is easy to use as no starter solution is required. The recommended dilution for both the tank and replenisher solution is 1+4. It is a robust, economical, clean working developer with a long tank life. It can be used at a wide range of developing temperatures, 20 – 40°C (68 – 104°F). With most papers it gives a print image tone just slightly cool of neutral.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty