Would you like to log in again?
Ilford 2000RT Machine Fixer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
83066-05
101381-668EA 143.43 USD InStock
101381-668
Ilford 2000RT Machine Fixer, Electron Microscopy Sciences
Film Processing Supplies
The 2000RT is a non-hardening, liquid fixer concentrate specially designed for the rapid fixing of resin coated paper in processing machines complementing the 2000RT developer. It can be used at a wide range of fixer temperatures, 20 – 40°C (68 – 104°F). 2000RT Fixer must not be used with fix hardeners.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty