Would you like to log in again?
Ilford Hypam Fixer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
83062-50
101381-666EA 73.83 USD InStock
101381-666
Ilford Hypam Fixer, Electron Microscopy Sciences
Film Processing Supplies
HYPAM is a non-hardening liquid rapid fixer that can be used with the fix hardening agents that are needed for some film processing applications. Without the addition of a hardener it is suitable for all black and white film and paper, manual and machine processing applications in the temperature range of 18 – 40°C (64 -104°F). HYPAM is the recommended fixer for all ILFORD scientific products and other X-ray films, plates and cineradiography films. If the addition of hardener is required, use ILFORD Hypam Hardener.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty