Would you like to log in again?
Wood Handle Metal Brush, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
65585-B 65585-S
101413-026EA 24.17 USD InStock
101413-026 101413-028
Wood Handle Metal Brush, Electron Microscopy Sciences
Brushes
A four-row brush made in Germany from 0.10 mm crimped steel or brass wire set in a wooden handle.
Excellent for use as a cleaning brush or for surface texturing on soft metals. The overall length is 8 5/8”.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty