Would you like to log in again?
Oxide Remover Cleaner, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71052 71053
101413-356EA 39.34 USD InStock
101413-356 101413-358
Oxide Remover Cleaner, Electron Microscopy Sciences
Cleaning Agents Detergents and Surface Cleansers
This cleaner may be used on brass, copper, steel and other metals to remove rust and metal-oxides.
  • Comes as a concentrate
Dilute to 10 to 12% with water for use with ultrasonic machine.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty