Would you like to log in again?
Flexible Spatulas, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63321
101412-830EA 32.51 USD InStock
101412-830
Flexible Spatulas, Electron Microscopy Sciences
Spatulas
This is a sword type spatula, made from stainless steel with a wooden handle.
  • Very thin
  • Has bendability toward the end and very stiff towards the handle
  • Overall length is 6³/₄"
It offers flexibility when working with soft materials, such as embedding waxes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty