Would you like to log in again?
INFINITYX-32 High Resolution CCD Cameras, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Infinity
95117 95116
102091-470EA 9648.95 USD InStock
102091-470 102091-468
INFINITYX-32 High Resolution CCD Cameras, Electron Microscopy Sciences
Cameras
The INFINITYX-32 is the first pixel shifting camera to utilize the highly sensitive SONY ICS274 sensor.
  • 32 megapixel pixel shifting camera
  • 2, 8, 16 and 32 megapixel resolution on capture
  • Highly sensitive, low noise CCD with remarkable color quality for the most demanding applications
  • 12 fps at full 1616X1216 resolution, 25 fps at smaller regions of interest
  • 8 & 12-bit selectable output format
Using patented pixel shifting technology, each CCD pixel captures full chromatic information resulting in increased color resolution with no degradation. Available at a fraction of the cost of competitive cameras, this complete imaging solution is an affordable, extraordinary tool for professionals in clinical, life and material sciences where megapixel resolution, color accuracy and light sensivity are essential.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty