Would you like to log in again?
Chemically Inert Polyester Tags on Sheets, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Tough-Tags®
77520-Y 77522-W 77522-B 77526-W 77524-W 77522-G 77523-Y 77520-W 77528-W 77520-R 77529-W 77523-O 77537-W 77525-W 77521-R 77523-R 77538-W 77522-R 77523-6 77530-C 77527-W 77523-W 77522-Y 77522-6 77531-C 77523-B 77520-O 77521-G 77521-O 77532-C 77521-W 77522-O 77520-G 77521-6 77521-Y 77520-6 77536-W 77521-B 77523-G 77520-B
101413-780PK 109.87 USD InStock
101413-780 101413-762 101413-784 101413-782 101413-760 101413-788 101413-766 101413-822 101413-800 101413-786 101413-764 101413-820 101413-826 101413-804 101413-768 101413-824 101413-802 101413-808 101413-806 101413-828 101413-770 101413-792 101413-790 101413-796 101413-774 101413-794 101413-772 101413-810 101413-778 101413-830 101413-798 101413-776 101413-814 101413-836 101413-812 101413-834 101413-758 101413-818 101413-816 101413-838
Chemically Inert Polyester Tags on Sheets, Electron Microscopy Sciences
Labels
Tough-Tags® labels are mainly used for microcentrifuge tubes and other containers. These labels are made of chemically inert polyester sheets which resist most organic solvents and caustic agents.
  • Adhere to most plastics, glass and metals without cracking, peeling or degrading
  • Temperature resistance: −40 to 121 °C
Label sheets are specially top-coated to readily accept laser printing. Stay-flat sheets will not jam in laser printers. Tubes labeled with Tough-Tags® are easily slide in and out of centrifuge rotors without binding.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty