Would you like to log in again?
Wax Stripper, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
50392-01
101411-926EA 73.23 USD InStock
101411-926
Wax Stripper, Electron Microscopy Sciences
Stains Microscopy and staining reagents
WaxStrip™ is an environmentally safe powder compound prepared primarily for the removal of mounting waxes in semiconductor processing.
  • Environmentally safe
  • Powder consistency
  • 1 lb/bottle
WaxStrip™ is compatible for use on silicon, glass, iron, aluminum, zinc, brass, copper, magnesium and other materials.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty