Would you like to log in again?
Cambridge, LEO, Phillips 500, Cameca Mounts, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75181 75180
101413-616PK 284.88 USD InStock
101413-616 101413-618
Cambridge, LEO, Phillips 500, Cameca Mounts, Electron Microscopy Sciences
Specimen Mounting Supplies
Aluminum small, slotted head. Surface measures: 9.6 x 11.2 mm (3/8 x 7/16"). Pin 3.1 mm (1/8").
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty