Would you like to log in again?
SEM Mount Forceps, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
76805
101413-698EA 42.13 USD InStock
101413-698
SEM Mount Forceps, Electron Microscopy Sciences
Forceps
Made from stainless steel with a serrated handle and a guide pin.
The tip is bent at a 45° and formed into a ring, which has a diameter of 10 mm when fully closed. 150 mm long.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty