Would you like to log in again?
FreezGuard™ Alarm, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72092-10 72092-20
102097-862EA 295.07 USD InStock
102097-862 102097-864
FreezGuard™ Alarm, Electron Microscopy Sciences
Data Loggers
Alarm will Sound for Up to Three Days. Prevent costly, time consuming sample loss with this practical battery operated system.
  • The alarm can be reset manually
  • A beep will signal when the 9 V battery (not included) is low
The FreeGuard™ Alarm monitors freezers and coolers and detects any variance from temperatures between –20 °C and 13 °C (–5 °F to 55 °F), ±1 °C, which are preset by the user. The resonant alarm will continue to sound for up to three days if the temperature falls out of the pre-set range. Should the temperature correct itself, an LED will indicate that a variance has occurred. For added protection, and extra life, a second battery can be connected. The unit mounts with self-adhesive tape or screw. Unit comes with a 3 m probe lead, and measures W×D×H: 86×35×26 mm (35/16×11/2×51/16"). The weight without the battery is 144 g (5.1 oz.).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty