Would you like to log in again?
Liquid Wicks, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60160-F 60160-XF 60160-15 60160-M
102093-092PK 44.54 USD InStock
102093-092 102093-094 102093-096 102093-098
Liquid Wicks, Electron Microscopy Sciences
Swabs Swabs for Cleaning
Size 15 wicks fit the wicking aperture of MicroMounts, and are ideal for removing residual liquid from your crystal and mounts
Theses paper wicks are offer a  great deal of absorbency with its small tip that foam-tipped or cotton-tipped applicators can not substitute. These wicks are available sizes: 15, extra fine (XF), fine (F), and Medium (M).
Size 15 wicks fit the wicking aperture of MicroMounts, and are ideal for removing residual liquid from your crystal and mounts. Extra Fine (XF) are also ideal for removing liquid formed around your crystal. Size Medium (M) and Fine (F) wicks are ideal for cleaning MiroMount The thick end of these wicks fit into standard 0.7 mm mechanical pencil.
Each box contains 300, 28 mm long high quality, tapered paper wicks.
Each box contains 300, 28 mm long high quality, tapered paper wicks.
Extra Fine (XF) are also ideal for removing liquid formed around your crystal. Size Medium (M) and Fine (F) wicks are ideal for cleaning MiroMount The thick end of these wicks fit into standard 0.7 mm mechanical pencil.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty