Would you like to log in again?
Tough-Tag Color Coded Labels, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Tough-Tags®
77539-Y 77539-G 77539-B 77539-W 77539-R
101413-848EA 94.15 USD InStock
101413-848 101413-846 101413-840 101413-844 101413-842
Tough-Tag Color Coded Labels, Electron Microscopy Sciences
Labels
Color coded tough tags will not come off in rotors, cyclers or racks.
  • Ideal for use on 1.5 ml tubes
  • Spot size is 0.44" (11 mm)
They are resistant to extreme temperatures and the matte surface is suitable for pencil, pen or marker.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty