Would you like to log in again?
Rocker Tape, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77512-20
102098-472RL 66.18 USD InStock
102098-472
Rocker Tape, Electron Microscopy Sciences
Tapes
Convenient - No more fumbling with clamps
Double-sided adhesive film for laboratory container mounting.
Convenient - No more fumbling with clamps
Versatile - Use with rockers, shakers, incubators or nutators
Strong - Easily rock and roll up to 300 rpm and beyond*
Long Lasting - Re-usable based on load capacity and vessel number
Resistent - Keeps on rockin’ even in the prescence of most solvents and caustic agents
* RPM limits may vary based on load and vessel contact area. Test load recommended
* RPM limits may vary based on load and vessel contact area. Test load recommended
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty