Would you like to log in again?
Drug Test Kit Mailer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61514-00
102093-432PK 120.65 USD InStock
102093-432
Drug Test Kit Mailer, Electron Microscopy Sciences
Mailers and Shippers
All in one, easy to use drug test mailing system that meets postal and courier service regulations for non-hazardous clinical specimens.
Model 600 kit consists of:
90 mL Specimen Bottle, Screw Top Cap and Temperature Indication Label 1 3/4” diameter
Pad of Absorbent Material 4 x 2 3/4”
Press-Lock Plastic Bag:, 5 7/8 x 9 3/8”
Model 601, I.D. 6 x 3 x 2 1/2”; O.D. 7 x 4 x 3 1/2”, 1/2” Wall
Model 602, I.D. 6 x 3 x 2 1/2”; O.D. 7 x 4 x 3 1/2”, 1/2” Wall
Security Seal: 7 x 1”
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty