Would you like to log in again?
Xylene Resistant Thermal Transfer Labels, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77598-10 77598-20
102098-738RL 126.66 USD InStock
102098-738 102098-740
Xylene Resistant Thermal Transfer Labels, Electron Microscopy Sciences
Labels
These Polyester labels provide strong solvent resistance, specifically to multiple xylene washes as well as longer term exposure to xylene.
  • Temperature range: −40 to 121 °C
  • Versatile and wide temperature applications
  • Top-coated to readily accept thermal transfer printing
These pre-cut, peel-off labels are available in a microscope slide size as well as a unique general-purpose size for multiple uses.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty