Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Metallurgical Microscope, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
BC-233
102099-948EA 3541.01 USD InStock
102099-948
Metallurgical Microscope, Electron Microscopy Sciences
Microscopes
For materials analysis and more
Optical body with Seidentopf design inclined 30°; 55 to 75 mm interpupillary adjustment, 360° rotation. Quintuple nosepiece with ball bearing. Eye pieces are 10X, with 25 mm focal Length and FN 18 mm. The focus mechanism has adjustable tension control to prohibit drift and adjustable up-stop to protect objective lenses; dial markings at 0.002mm increments. Incident light system includes condenser, iris diaphragm, color and polarizing filter. Transmitted illumination, 1.25 N.A. Abbe condenser with iris diaphragm; all systems use a 20 watt, 6 volt Halogen bulb with dimmer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty