Would you like to log in again?
Fluoro Gel with DABCO, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
Fluoro-Gel
17985-04 17985-02 17985-01 17985-03
102092-108EA 630 USD InStock
102092-108 102092-106 102092-110 102092-104
Fluoro Gel with DABCO, Electron Microscopy Science
Stains Microscopy and staining reagents
  • Ready to use mounting medium with anti-fading agent
  • Use in Immuno-fluorescence, confocol microscopy.
Fluoro Gel with antifading agent 1, 4-Diazobicyclo-(2,2,2-octane (DABCO™) is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Alexa fluoro 488, Alexa fluoro 594, Cy dyes, tetramethyly rhodamine, and Redox. The fluorescence is retained during prolonged storage at 4°C in the dark.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty