Would you like to log in again?
EMS Shield Mount , Electron Microscopy Science
Supplier: Electron Microscopy Sciences
17985-09 17985-100
102092-120EA 137.51 USD InStock
102092-120 102092-124
EMS Shield Mount , Electron Microscopy Science
Stains Microscopy and staining reagents
EMS Shield Mount  is an aqueous non-permanent, low viscosity mounting medium for immunofluorescence. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa fluoro 488, Alexa fluoro 594, Green fluorescent protein (GFP), tetramethyly rhodamine and Redox. The fluorescence is retained during storage at 4°C in the dark.
  • Ready to use mounting medium for Immunofluorescence
  • Does not contain phenylenediamine, which destroys immunofluorescence of Cy dyes
  • Use in Immuno-fluorescence, confocol microscopy.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty