Would you like to log in again?
DuraSeal Cling, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70994-07
102681-200EA 46.9 USD InStock
102681-200
DuraSeal Cling, Electron Microscopy Sciences
Seals General Purpose Sealing Films
Film can cling to itself and glass vessels
Heat Resistant, softens at 100°C. Strong film that can streth 300-400% before breaking or tearing. Tight Sealing, forms and air tight barrier when stretched across openings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty