Would you like to log in again?
Hydromount™ Nonfluorescing Mounting Media, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
Hydromount™
17966
102090-992EA 143.15 USD InStock
102090-992
Hydromount™ Nonfluorescing Mounting Media, Electron Microscopy Science
Stains Microscopy and staining reagents
Hydromount™ is the traditional choice whenever a nonfluorescing aqueous medium is needed. Hydromount™ is water-based and is suitable for mounting specimens that have been processed in water. Hydromount™ is effective for frozen sections, amyloid, and immunofluorescent staining procedures. Should it become necessary, Hydromount™ may be removed by soaking the slides in warm saline.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty