Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PEEK Polish, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
PEEK
71833-12 71833-13 71833-14 71833-11 71833-10
102097-692EA 24.39 USD InStock
102097-692 102681-202 102097-696 102097-698 102097-700
PEEK Polish, Electron Microscopy Sciences
Cleaning Agents Detergents and Surface Cleansers
PEEK is a complete line of cleaning materials that not only cleans but polishes and protects all metals as well as fiberglass surfaces as well. It is available in 5 different packaging configurations.

  • Boiling point: 175 °C (347 °F)
  • Flash point: 56 °C (133 °F)
  • Density: 1.03 g/cm³ (8.595 lbs/gal)

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty