Would you like to log in again?
Formalin Prefilled HistoTainer, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
15743-60 15743-120 15743-90 15743-40 15743-20
102091-914PK 60.55 USD InStock
102091-914 102091-916 102091-910 102091-912 102091-908
Formalin Prefilled HistoTainer, Electron Microscopy Science
Stains Microscopy and staining reagents
The HistoTainer™ (Empty Vials, see page 482) is half filled with 10% Neutral Buffered Formalin as a fixative. These ready-to-use prefilled containers are timer saving and make your work more efficient.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty