Would you like to log in again?
Large Paraffin Wax Dispenser, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62840-20 62840-10
102094-474EA 5619.88 USD InStock
102094-474 102094-472
Large Paraffin Wax Dispenser, Electron Microscopy Sciences
Paraffin Wax Trimmers and Dispensers
  • Microprocessor controlled – Accurate and uniformity temperature control
  • Security – Advanced software and mechanical devices assure safety
Liquid Paraffin Wax Dispenser holds 6.25 gallons of paraffin wax with clear lexan lid permits easy viewing of paraffin without contamination. High spigot accommodates large transfer vessels. The unit comes with microprocessor temperature regulation, built-in clock. Ambient to 65°C. Dimensions W x D x H: 35.6 x 38.1 x 58.4 cm (14 x 15 x 23"). Weight 20.5 kg (45 lbs.).
The Paraffin Dispenser have been tested by a registered independent body to rigorous international quality and safety standards, and, have been found to be in compliance with UL, CSA and CE.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty