Would you like to log in again?
Extra Wide Lab Cart, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64383-20 64383-60
102095-212EA 2267.1 USD InStock
102095-212 102095-208
Extra Wide Lab Cart, Electron Microscopy Sciences
Carts
Large storage area with extra wide drawers and compartments, heavy-duty casters with two locking.
  • HDPE top and bottom
  • Built-in handle for easy handling
  • 1 large compartment with hinged acrylic door
  • 3 extra-wide drawers to store supplies
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty