Would you like to log in again?
Refrigerator Storage & Trays, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64361-10
102095-092EA 743.02 USD InStock
102095-092
Refrigerator Storage & Trays, Electron Microscopy Sciences
Trays/Dishes/Pots Instrument Trays/Dishes
These storage trays designed to fit in a refrigerator or freezer.
  • Adjustable dividers (color Blue)
  • Ventilated designed to allow air movement
  • Diver can be order separately
  • Measures: 15.5L×15.8W×8.5"H
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty