Would you like to log in again?
NBS 25 Resolution Chart, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68098-10
102096-176EA 381.03 USD InStock
102096-176
NBS 25 Resolution Chart, Electron Microscopy Sciences
Charts
This consists of elements of 5 bars ranging from 1 to 18 lines per mm, with adjacent numbers indicating the number of lines per mm.
  • 1 to 18 lines per mm Chrome Surface Image
Produced as a vacuum deposited chrome image on 75 x 75 mm glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty