Would you like to log in again?
Digital Dry Bath, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
67119-30 67119-20 67119-10 67119-40
102095-990EA 1627.11 USD InStock
102095-990 102095-992 102095-988 102095-994
Digital Dry Bath, Electron Microscopy Sciences
Block Heaters Digital Block Heaters
Thermally control samples during use.
  • Utilizes all CoolSafe system chambers
  • Temperatures can be controlled from 5° above ambient to 100°C in 1° increments
  • Aluminum block evenly transfer heat to tubes
  • Each unit comes with a block lifter to safely remove hot chambers
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty