Would you like to log in again?
DiATOME® Ultratrim 45 Trimming Tool, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
DUTB45-R
102681-520EA 1447.47 USD InStock
102681-520
DiATOME® Ultratrim 45 Trimming Tool, Electron Microscopy Sciences
Knives
Diatome Ultratrim allows quick, easy and accurate trimming at both room and cryo-temperatures.
Due to the extreme sharpness, less mechanical damage is applied to the sample during trimming.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty