Would you like to log in again?
Wafer-Mount™ 559, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
Wafer-Mount™
50403-02 50403-10
100490-020PK 268.34 USD InStock
100490-020 100490-018
Wafer-Mount™ 559, Electron Microscopy Science
Stains Microscopy and staining reagents
Available in sheet form 10"x10" (254x254mm). To use, remove the paper backing exposing the adhesive layer.
The substrate is positioned on the film and pressed down firmly so that it adheres to the plastic film. No heating is required. The wafer mounted on the plastic film can then be held down using a vacuum manifold or mechanical means for dicing or scribing. After dicing or scribing, the parts can be separated from the wafer-mount 559 film by washing in acetone or MEK. Ideal for holding down silicon wafers, alumina, or glass substrates for scribing.
Sheet 10"x10" (254x254mm). Press down firmly to adhere substrate to film. No heating is required. Ideal for holding down silicon wafers, alumina, or glass substrates for scribing. Dissolves in acetone or MEK
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty