Would you like to log in again?
Omniprobe Copper 5-Post Lift-Out Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75964-04
102681-296PK 380.82 USD InStock
102681-296
Omniprobe Copper 5-Post Lift-Out Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
Custom copper 5-post lift-out grids specially designed for in-situ lift-out. These grids include multiple indexed mounting locations, all with vertical bars attachment surfaces. Now with lower profile sides for easier access to outermost posts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty